• USA Home
  • 81655 - O-tert-Butyl-L-threonine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
81655 Sigma-Aldrich

O-tert-Butyl-L-threonine methyl ester hydrochloride

≥98.0% (TLC)

  • CAS Number 71989-43-0

  • Empirical Formula (Hill Notation) C9H19NO3 · HCl

  • Molecular Weight 225.71

  •  Beilstein/REAXYS Number 4716005

  •  MDL number MFCD00077111

  •  PubChem Substance ID 57652884

  •  NACRES NA.22

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Threonine More...
InChI Key   SDHKEUUZUMQSAD-HHQFNNIRSA-N
assay   ≥98.0% (TLC)
storage temp.   2-8°C

Description

Application

O-tert-Butyl-L-threonine methyl ester hydrochloride may be used in the preparation of N-acryl-O-tert-butyl methyl ester-L-threonine, which may undergo miniemulsion polymerization to form the corresponding chiral polymer nanoparticles with potential application in enantioselective crystallization.

Packaging

5 g in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?