EMAIL THIS PAGE TO A FRIEND
84607 Sigma-Aldrich

SCAL-Linker

technical, ≥90% (HPLC)

Synonym: 4,4′-Bis(methylsulfinyl)-2-(4-carboxybutoxy)-N-Fmoc-benzhydrylamine, Safety catch amide linker

Purchase

Properties

grade   technical
assay   ≥90% (HPLC)
storage temp.   2-8°C
SMILES string   CS(=O)c1ccc(cc1)C(NC(=O)OCC2c3ccccc3-c4ccccc24)c5ccc(cc5OCCCCC(O)=O)S(C)=O

Description

Other Notes

Safety-catch anchoring linker for solid-phase peptide synthesis

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?