• USA Home
  • 84985 - L-Serine β-lactone tetrafluoroborate salt

EMAIL THIS PAGE TO A FRIEND
84985 Sigma-Aldrich

L-Serine β-lactone tetrafluoroborate salt

≥98.0% (T)

Synonym: (S)-3-Amino-2-oxetanone tetrafluoroborate salt

  • CAS Number 403655-97-0

  • Empirical Formula (Hill Notation) C3H6NO2 · BF4

  • Molecular Weight 174.89

  •  MDL number MFCD01863773

  •  PubChem Substance ID 57653030

Purchase

Properties

Related Categories Amino Acids & Derivatives, Asymmetric Synthesis, Building Blocks, Chemical Biology, Chemical Synthesis,
InChI Key   ZYHZSWQYHKJXBX-DKWTVANSSA-O
assay   ≥98.0% (T)
optical activity   [α]20/D −17±2°, c = 1% in DMF
mp   ~150 °C (dec.)
storage temp.   2-8°C

Description

Other Notes

Versatile building block for the synthesis of unnatural (S)-α-amino acids (3-substituted L-alanines)

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Articles

Oxetanes

Oxetanes are the closest homologs to epoxides, but historically have received far less attention than their three-membered ringed brethren. However, oxetanes have received increasing exposure as attr...
Mark Redlich
Chemfiles Volume 10 Article 4
Keywords: Antibiotics, Building blocks, Drug discovery, Medicinal chemistry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?