• USA Home
  • 850225 - 3,5-Diiodo-L-tyrosine dihydrate

EMAIL THIS PAGE TO A FRIEND
850225 Sigma-Aldrich

3,5-Diiodo-L-tyrosine dihydrate

98%

  • CAS Number 18835-59-1

  • Empirical Formula (Hill Notation) C9H9I2NO3 · 2H2O

  • Molecular Weight 469.01

  •  Beilstein/REAXYS Number 2218691

  •  MDL number MFCD00016542

Purchase

Properties

InChI Key   NYPYHUZRZVSYKL-ZETCQYMHSA-N
assay   98%
optical activity   [α]20/D +1.5°, c = 4 in 1 M HCl
mp   200 °C (dec.) (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?