• USA Home
  • 850373 - D-(+)-Melezitose hydrate

EMAIL THIS PAGE TO A FRIEND
850373 Sigma-Aldrich

D-(+)-Melezitose hydrate

Synonym: α-D-Glc-[1→3]-β-D-Fru-[2→1]-α-D-Glc

  • CAS Number 207511-10-2

  • Empirical Formula (Hill Notation) C18H32O16 · xH2O

  • Molecular Weight 504.44 (anhydrous basis)

  •  EC Number 209-894-9

  •  MDL number MFCD00149448

Purchase

Properties

InChI Key   CFXAVQUXSYFPDE-ZDVKUAOASA-N
optical activity   [α]23/D +85.9°, c = 4 in H2O
mp   160 °C (dec.) (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

M5375 D-(+)-Melezitose hydrate, ≥97% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?