• USA Home
  • 851159 - N-Chloroacetyl-L-tyrosine

EMAIL THIS PAGE TO A FRIEND
851159 Sigma-Aldrich

N-Chloroacetyl-L-tyrosine

98%

  • CAS Number 1145-56-8

  • Empirical Formula (Hill Notation) C11H12ClNO4

  • Molecular Weight 257.67

  •  MDL number MFCD00002390

Purchase

Properties

assay   98%
SMILES string   OC(=O)C(Cc1ccc(O)cc1)NC(=O)CCl
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?