• USA Home
  • 851213 - S-Carboxymethyl-L-cysteine

EMAIL THIS PAGE TO A FRIEND
851213 Aldrich

S-Carboxymethyl-L-cysteine

98%

Synonym: 3-carboxymethylthio-L-alanine

  • CAS Number 638-23-3

  • Empirical Formula (Hill Notation) C5H9NO4S

  • Molecular Weight 179.19

  •  Beilstein/REAXYS Number 1725012

  •  MDL number MFCD00002614

  •  PubChem Substance ID 329769357

Purchase

Properties

InChI Key   GBFLZEXEOZUWRN-VKHMYHEASA-N
assay   98%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C7757 S-Carboxymethyl-L-cysteine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?