• USA Home
  • 851892 - L-Cysteic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
851892 Sigma-Aldrich

L-Cysteic acid monohydrate

98%

Synonym: (R)-2-Amino-3-sulfopropionic acid

  • CAS Number 23537-25-9

  • Linear Formula HO3SCH2CH(NH2)CO2H·H2O

  • Molecular Weight 187.17

  •  Beilstein/REAXYS Number 3714036

  •  EC Number 207-861-3

  •  MDL number MFCD00149544

Purchase

Properties

assay   98%
optical activity   [α]24/D +8.2°, c = 7.5 in H2O
mp   267 °C (dec.) (lit.)
SMILES string   [H]O[H].N[C@@H](CS(O)(=O)=O)C(O)=O
InChI   1S/C3H7NO5S.H2O/c4-2(3(5)6)1-10(7,8)9;/h2H,1,4H2,(H,5,6)(H,7,8,9);1H2/t2-;/m0./s1
InChI key   PCPIXZZGBZWHJO-DKWTVANSSA-N
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

30170 L-Cysteic acid monohydrate, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?