EMAIL THIS PAGE TO A FRIEND
851914 Sigma-Aldrich

3-Nitro-L-tyrosine

99%

  • CAS Number 621-44-3

  • Linear Formula O2NC6H3-4-(OH)CH2CH(NH2)CO2H

  • Molecular Weight 226.19

  •  Beilstein/REAXYS Number 2813157

  •  EC Number 210-688-6

  •  MDL number MFCD00007123

  •  eCl@ss 32160406

Purchase

Properties

assay   99%
optical activity   [α]22/D +3.8°, c = 1 in 1 M HCl
mp   233-235 °C (dec.) (lit.)
SMILES string   N[C@@H](Cc1ccc(O)c(c1)[N+]([O-])=O)C(O)=O
InChI   1S/C9H10N2O5/c10-6(9(13)14)3-5-1-2-8(12)7(4-5)11(15)16/h1-2,4,6,12H,3,10H2,(H,13,14)/t6-/m0/s1
InChI key   FBTSQILOGYXGMD-LURJTMIESA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N7389 3-Nitro-L-tyrosine, crystalline

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?