• USA Home
  • 852201 - 2′-Deoxycytidine 5′-monophosphate

EMAIL THIS PAGE TO A FRIEND
852201 Sigma-Aldrich

2′-Deoxycytidine 5′-monophosphate

98%

Synonym: Deoxycytidylic acid, dCMP

  • CAS Number 1032-65-1

  • Empirical Formula (Hill Notation) C9H14N3O7P

  • Molecular Weight 307.20

  •  Beilstein/REAXYS Number 41062

  •  EC Number 213-849-9

  •  MDL number MFCD00006546

Purchase

Properties

assay   98%
mp   179-180 °C (dec.) (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

D7750 2′-Deoxycytidine 5′-monophosphate, Sigma Grade, ≥95.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?