EMAIL THIS PAGE TO A FRIEND
855243 Aldrich

Creatine hydrate

99%

Synonym: (α-Methylguanido)acetic acid, N-Amidinosarcosine, N-Guanyl-N-methylglycine

  • Linear Formula H2NC(=NH)N(CH3)CH2CO2H · xH2O

  • Molecular Weight 131.13 (anhydrous basis)

  •  EC Number 200-306-6

  •  MDL number MFCD00071582

Purchase

Properties

assay   99%
mp   292 °C (dec.) (lit.)
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
MB7706000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C3630 Creatine monohydrate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?