• USA Home
  • 855502 - 3-O-Methyl-D-glucopyranose

EMAIL THIS PAGE TO A FRIEND
855502 Sigma-Aldrich

3-O-Methyl-D-glucopyranose

97%

Synonym: 3-O-Methyl-α-D-glucopyranose, 3-O-Methylglucose

Purchase

Properties

InChI Key   SCBBSJMAPKXHAH-OVHBTUCOSA-N
assay   97%
optical activity   [α]20/D +104.5°, c = 1 in H2O
mp   167-169 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M4879 3-O-Methyl-D-glucopyranose, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?