EMAIL THIS PAGE TO A FRIEND
856290 Sigma-Aldrich

Sterigmatocystin

Synonym: 3a,12c-Dihydro-8-hydroxy-6-methoxy-7H-furol[3′,2′:4,5]furo[2,3-c]xanthen-7-one

  • CAS Number 10048-13-2

  • Empirical Formula (Hill Notation) C18H12O6

  • Molecular Weight 324.28

  •  Beilstein/REAXYS Number 53259

  •  EC Number 233-158-6

  •  MDL number MFCD00005084

Purchase

Properties

SMILES string   COc1cc2OC3OC=CC3c2c4Oc5cccc(O)c5C(=O)c14
InChI   1S/C18H12O6/c1-21-11-7-12-13(8-5-6-22-18(8)24-12)17-15(11)16(20)14-9(19)3-2-4-10(14)23-17/h2-8,18-19H,1H3
InChI key   UTSVPXMQSFGQTM-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Initiates lung and liver tumors under experimental conditions.

Features and Benefits

A mildly carcinogenic metabolite produced by several strains of Aspergillus.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGIII
WGK Germany 
3
RTECS 
LV1750000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

S3255 Sterigmatocystin, powder, ≥98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?