EMAIL THIS PAGE TO A FRIEND
856649 Sigma-Aldrich

L-(−)-Mannose

98%

Synonym: L-Mannopyranose

  • CAS Number 10030-80-5

  • Empirical Formula (Hill Notation) C6H12O6

  • Molecular Weight 180.16

  •  Beilstein/REAXYS Number 1724628

  •  EC Number 233-080-2

  •  MDL number MFCD00063884

Purchase

Properties

assay   98%
optical activity   [α]24/D −14.3°, c = 4 in H2O
mp   129-131 °C (lit.)
SMILES string   OC[C@@H]1OC(O)[C@H](O)[C@H](O)[C@H]1O
InChI   1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m0/s1
InChI key   WQZGKKKJIJFFOK-JFNONXLTSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

M1134 L-(−)-Mannose, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?