EMAIL THIS PAGE TO A FRIEND
856878 Aldrich

Ala-Gly

Synonym: L-Alanyl-glycine

  • CAS Number 687-69-4

  • Linear Formula CH3CH(NH2)CONHCH2COOH

  • Molecular Weight 146.14

  •  Beilstein Registry Number 1723438

  •  MDL number MFCD00065107

  •  PubChem Substance ID 329769403

Purchase

Properties

InChI Key   CXISPYVYMQWFLE-VKHMYHEASA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?