• USA Home
  • 857793 - Cytochalasin A from Helminthosporium dematioideum

EMAIL THIS PAGE TO A FRIEND
857793 Aldrich

Cytochalasin A from Helminthosporium dematioideum

  • CAS Number 14110-64-6

  • Empirical Formula (Hill Notation) C29H35NO5

  • Molecular Weight 477.59

  •  Beilstein Registry Number 949521

  •  EC Number 237-964-9

  •  MDL number MFCD00005935

  •  PubChem Substance ID 329769413

Purchase

Properties

InChI Key   ZMAODHOXRBLOQO-TZVKRXPSSA-N

Description

Features and Benefits

The cytochalasins are fungal metabolites which exhibit interesting effects on cell activity. They are important tools for cytological research.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?