EMAIL THIS PAGE TO A FRIEND
858471 Sigma-Aldrich

5-Fluorouracil

99%

Synonym: 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione

  • CAS Number 51-21-8

  • Empirical Formula (Hill Notation) C4H3FN2O2

  • Molecular Weight 130.08

  •  Beilstein/REAXYS Number 127172

  •  EC Number 200-085-6

  •  MDL number MFCD00006018

  •  PubChem Substance ID 24888466

Purchase

Properties

assay   99%
mp   282-286 °C (dec.) (lit.)
solubility   ammonium hydroxide: 50 mg/mL, clear, colorless
SMILES string   FC1=CNC(=O)NC1=O
InChI   1S/C4H3FN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9)
InChI key   GHASVSINZRGABV-UHFFFAOYSA-N
Gene Information   human ... TYMS(7298)

Description

Application

Inhibitor of DNA synthesis.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

F6627 5-Fluorouracil, ≥99% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?