EMAIL THIS PAGE TO A FRIEND
860875 Sigma-Aldrich

Brilliant Green

certified by the Biological Stain Commission

Synonym: Astradiamant green GX, Basic Green 1, Diamond Green, Emerald Green, Ethyl Green, Malachite Green G, Solid Green JO

  • CAS Number 633-03-4

  • Empirical Formula (Hill Notation) C27H34N2O4S

  • Molecular Weight 482.63

  •  Colour Index Number 42040

  •  Beilstein/REAXYS Number 3901207

  •  EC Number 211-190-1

  •  MDL number MFCD00011880

Purchase

Properties

grade   certified by the Biological Stain Commission
InChI Key   NNBFNNNWANBMTI-UHFFFAOYSA-M
composition   , ~90%
mp   210 °C (dec.) (lit.)
λmax   428 nm
  625 nm (2nd)
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
BP6825000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

B4014 Brilliant Green, certified by the Biological Stain Commission

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?