• USA Home
  • 861596 - 4-[N-(2-Amino-3-cyano-5-pyrazinylmethyl)amino]benzoic acid

EMAIL THIS PAGE TO A FRIEND
861596 Sigma-Aldrich

4-[N-(2-Amino-3-cyano-5-pyrazinylmethyl)amino]benzoic acid

98%

  • CAS Number 100900-25-2

  • Empirical Formula (Hill Notation) C13H11N5O2

  • Molecular Weight 269.26

  •  MDL number MFCD00006138

Purchase

Properties

assay   98%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?