• USA Home
  • 88412 - cis-4-(Boc-amino)cyclohexanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
88412 Sigma-Aldrich

cis-4-(Boc-amino)cyclohexanecarboxylic acid

≥98.0% (TLC)

  • CAS Number 53292-90-3

  • Linear Formula (CH3)3CO2CNHC6H10CO2H

  • Molecular Weight 243.30

  •  Beilstein Registry Number 7641071

  •  MDL number MFCD03453262

  •  PubChem Substance ID 57653302

Purchase

Properties

Related Categories Alicyclic Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Chemistry | Peptide Synthesis, Unnatural Amino Acids & Derivatives More...
InChI Key   KXMRDHPZQHAXML-UHFFFAOYSA-N
assay   ≥98.0% (TLC)

Description

Packaging

500 mg in poly bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?