• USA Home
  • 88726 - (S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldi(3,5-xylyl)phosphine

EMAIL THIS PAGE TO A FRIEND
88726 Sigma-Aldrich

(S)-1-[(RP)-2-(Diphenylphosphino)ferrocenyl]ethyldi(3,5-xylyl)phosphine

≥97%

Synonym: (2S)-1-[(1S)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(diphenylphosphino)ferrocene (acc to CAS), Josiphos SL-J005-2

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Josiphos Ligands, Privileged Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)P(c5cc(C)cc(C)c5)c6cc(C)cc(C)c6
InChI   1S/C35H35P2.C5H5.Fe/c1-25-19-26(2)22-32(21-25)36(33-23-27(3)20-28(4)24-33)29(5)34-17-12-18-35(34)37(30-13-8-6-9-14-30)31-15-10-7-11-16-31;1-2-4-5-3-1;/h6-24,29H,1-5H3;1-5H;/t29-;;/m0../s1
InChI key   VIWMSTLBMCHMGT-UJXPALLWSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?