• USA Home
  • 88741 - (R)-1-{(SP)-2-[Di(2-furyl)phosphino]ferrocenyl}ethyldi-tert-butylphosphine

EMAIL THIS PAGE TO A FRIEND
88741 Sigma-Aldrich

(R)-1-{(SP)-2-[Di(2-furyl)phosphino]ferrocenyl}ethyldi-tert-butylphosphine

≥97%

Synonym: (2R)-1-[(1R)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene (acc to CAS), Josiphos SL-J212-1

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Josiphos Ligands, Privileged Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(c3ccco3)c4ccco4)P(C(C)(C)C)C(C)(C)C
InChI   1S/C23H31O2P2.C5H5.Fe/c1-17(27(22(2,3)4)23(5,6)7)18-11-8-12-19(18)26(20-13-9-15-24-20)21-14-10-16-25-21;1-2-4-5-3-1;/h8-17H,1-7H3;1-5H;/t17-;;/m1../s1
InChI key   CVBADLBJGBTJDF-ZEECNFPPSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Solvias Chiral Phosphine Ligands - Toolkit for Asymmetric Catalysis

Sigma-Aldrich, in collaboration with Solvias, is proud to present the Chiral Ligands Kit – the ultimate toolkit for asymmetric catalysis!
Keywords: Asymmetric catalysis, Asymmetric synthesis, Catalysis, Individual protein Immunoprecipitation, Ligands

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?