• USA Home
  • 88742 - (S)-1-{(RP)-2-[Di(2-furyl)phosphino]ferrocenyl}ethyldi-tert-butylphosphine

EMAIL THIS PAGE TO A FRIEND
88742 Sigma-Aldrich

(S)-1-{(RP)-2-[Di(2-furyl)phosphino]ferrocenyl}ethyldi-tert-butylphosphine

≥97%

Synonym: (2S)-1-[(1S)-1-[Bis(1,1-dimethylethyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene (acc to CAS), Josiphos SL-J212-2

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Josiphos Ligands, Privileged Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2P(c3ccco3)c4ccco4)P(C(C)(C)C)C(C)(C)C
InChI   1S/C23H31O2P2.C5H5.Fe/c1-17(27(22(2,3)4)23(5,6)7)18-11-8-12-19(18)26(20-13-9-15-24-20)21-14-10-16-25-21;1-2-4-5-3-1;/h8-17H,1-7H3;1-5H;/t17-;;/m0../s1
InChI key   CVBADLBJGBTJDF-RMRYJAPISA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?