• USA Home
  • 88745 - (R)-1-{(SP)-2-[Di(1-naphthyl)phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine

EMAIL THIS PAGE TO A FRIEND
88745 Sigma-Aldrich

(R)-1-{(SP)-2-[Di(1-naphthyl)phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine

≥97%

Synonym: (2R)-1-[(1R)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-(di-1-naphthalenylphosphino)ferrocene (acc to CAS), Josiphos SL-J404-1

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Josiphos Ligands, Privileged Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(c3cccc4ccccc34)c5cccc6ccccc56)P(c7cc(C)cc(C)c7)c8cc(C)cc(C)c8
InChI   1S/C43H39P2.C5H5.Fe/c1-29-23-30(2)26-36(25-29)44(37-27-31(3)24-32(4)28-37)33(5)38-19-12-22-41(38)45(42-20-10-15-34-13-6-8-17-39(34)42)43-21-11-16-35-14-7-9-18-40(35)43;1-2-4-5-3-1;/h6-28,33H,1-5H3;1-5H;/t33-;;/m1../s1
InChI key   NODPGYHIGOCWSG-QYKZUBHNSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Solvias Chiral Phosphine Ligands - Toolkit for Asymmetric Catalysis

Sigma-Aldrich, in collaboration with Solvias, is proud to present the Chiral Ligands Kit – the ultimate toolkit for asymmetric catalysis!
Keywords: Asymmetric catalysis, Asymmetric synthesis, Catalysis, Individual protein Immunoprecipitation, Ligands

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?