• USA Home
  • 88748 - (S)-1-{(RP)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethyldi(3,5-xylyl)phosphine

EMAIL THIS PAGE TO A FRIEND
88748 Sigma-Aldrich

(S)-1-{(RP)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethyldi(3,5-xylyl)phosphine

≥97%

Synonym: (2S)-1-[(1S)-1-[Bis(3,5-dimethylphenyl)phosphino]ethyl]-2-[bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocene (acc to CAS), Josiphos SL-J418-2

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Josiphos Ligands, Privileged Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.COc2c(C)cc(cc2C)P([C]3[CH][CH][CH][C]3[C@H](C)P(c4cc(C)cc(C)c4)c5cc(C)cc(C)c5)c6cc(C)c(OC)c(C)c6
InChI   1S/C41H47O2P2.C5H5.Fe/c1-25-15-26(2)18-34(17-25)44(35-19-27(3)16-28(4)20-35)33(9)38-13-12-14-39(38)45(36-21-29(5)40(42-10)30(6)22-36)37-23-31(7)41(43-11)32(8)24-37;1-2-4-5-3-1;/h12-24,33H,1-11H3;1-5H;/t33-;;/m0../s1
InChI key   WMHNOIYFVOSUJD-NYPSMHOZSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?