• USA Home
  • 88749 - (R)-1-{(SP)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethylbis(2-methylphenyl)phosphine

EMAIL THIS PAGE TO A FRIEND
88749 Sigma-Aldrich

(R)-1-{(SP)-2-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]ferrocenyl}-ethylbis(2-methylphenyl)phosphine

≥97%

Synonym: (1R)-1-[Bis(4-methoxy-3,5-dimethylphenyl)phosphino]-2-[(1R)-1-[bis(2-methylphenyl)phosphino]ethyl]ferrocene (acc to CAS), Josiphos SL-J425-1

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Josiphos Ligands, Privileged Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.COc2c(C)cc(cc2C)P([C]3[CH][CH][CH][C]3[C@@H](C)P(c4ccccc4C)c5ccccc5C)c6cc(C)c(OC)c(C)c6
InChI   1S/C39H43O2P2.C5H5.Fe/c1-25-15-10-12-18-35(25)42(36-19-13-11-16-26(36)2)31(7)34-17-14-20-37(34)43(32-21-27(3)38(40-8)28(4)22-32)33-23-29(5)39(41-9)30(6)24-33;1-2-4-5-3-1;/h10-24,31H,1-9H3;1-5H;/t31-;;/m1../s1
InChI key   WONLWQAKBNPXTQ-OGLGXUNJSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Solvias Chiral Phosphine Ligands - Toolkit for Asymmetric Catalysis

Sigma-Aldrich, in collaboration with Solvias, is proud to present the Chiral Ligands Kit – the ultimate toolkit for asymmetric catalysis!
Keywords: Asymmetric catalysis, Asymmetric synthesis, Catalysis, Individual protein Immunoprecipitation, Ligands

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?