• USA Home
  • 88751 - (R)-1-{(SP)-2-[Di(2-furyl)phosphino]ferrocenyl}ethylbis(2-methylphenyl)phosphine

EMAIL THIS PAGE TO A FRIEND
88751 Sigma-Aldrich

(R)-1-{(SP)-2-[Di(2-furyl)phosphino]ferrocenyl}ethylbis(2-methylphenyl)phosphine

≥97%

Synonym: (2R)-1-[(1R)-1-[Bis(2-methylphenyl)phosphino]ethyl]-2-(di-2-furanylphosphino)ferrocene (acc to CAS), Josiphos SL-J452-1

  • CAS Number 849924-73-8

  • Empirical Formula (Hill Notation) C34H32FeO2P2

  • Molecular Weight 590.41

  •  MDL number MFCD08561168

  •  PubChem Substance ID 24889139

  •  NACRES NA.22

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Josiphos Ligands, Privileged Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
storage temp.   room temp
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(c3ccco3)c4ccco4)P(c5ccccc5C)c6ccccc6C
InChI   1S/C29H27O2P2.C5H5.Fe/c1-21-11-4-6-14-25(21)32(26-15-7-5-12-22(26)2)23(3)24-13-8-16-27(24)33(28-17-9-19-30-28)29-18-10-20-31-29;1-2-4-5-3-1;/h4-20,23H,1-3H3;1-5H;/t23-;;/m1../s1
InChI key   SPAUEXSFJREZBQ-MQWQBNKOSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Solvias Chiral Phosphine Ligands - Toolkit for Asymmetric Catalysis

Sigma-Aldrich, in collaboration with Solvias, is proud to present the Chiral Ligands Kit – the ultimate toolkit for asymmetric catalysis!
Keywords: Asymmetric catalysis, Asymmetric synthesis, Catalysis, Individual protein Immunoprecipitation, Ligands

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?