• USA Home
  • 89178 - (R)-Boc-3-cyano-β-Phe-OH

EMAIL THIS PAGE TO A FRIEND
89178 Sigma-Aldrich

(R)-Boc-3-cyano-β-Phe-OH

≥98.0% (HPLC)

Synonym: (R)-3-(Boc-amino)-3-(3-cyanophenyl)propionic acid, Boc-3-cyano-L-β-phenylalanine

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
Quality Level   100
assay   ≥98.0% (HPLC)
application(s)   peptide synthesis: suitable
SMILES string   CC(C)(C)OC(=O)N[C@H](CC(O)=O)c1cccc(c1)C#N
InChI   1S/C15H18N2O4/c1-15(2,3)21-14(20)17-12(8-13(18)19)11-6-4-5-10(7-11)9-16/h4-7,12H,8H2,1-3H3,(H,17,20)(H,18,19)/t12-/m1/s1
InChI key   QSPUAVCHVGCBGN-GFCCVEGCSA-N

Description

Packaging

500 mg in poly tube

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?