EMAIL THIS PAGE TO A FRIEND
89706 Sigma-Aldrich

Triethylsilane

produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (GC)

Synonym: NSC 93579, Silane E3H, Triethylhydrosilane, Triethylsilicon hydride

  • CAS Number 617-86-7

  • Linear Formula (C2H5)3SiH

  • Molecular Weight 116.28

  •  Beilstein/REAXYS Number 1098278

  •  EC Number 210-535-3

  •  MDL number MFCD00009018

  •  PubChem Substance ID 24889253

Purchase

Properties

grade   produced by Wacker Chemie AG, Burghausen, Germany
InChI Key   AQRLNPVMDITEJU-UHFFFAOYSA-N
assay   ≥98.0% (GC)
mfr. no.   Wacker Chemie AG
refractive index   n20/D 1.412 (lit.)
bp   107-108 °C(lit.)
density   0.728 g/mL at 25 °C (lit.)

Description

Application

Catalyst for:
• Synthesis of a spiro-oxindole blocker of Nav1.7 for the treatment of pain
• Redox initiated cationic polymerization
• Beckmann rearrangement of cyclododecanone oxime
• Regioselective reductive coupling of enones and allenes

Catalyst reactivation after catalyst polymerization of styrene

Studies involving the prediction of organosilicon flash points

Other Notes

Silane E3H

prices for bulk quantities on request

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
1
Flash Point(F) 
26.6 °F
Flash Point(C) 
-3 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?