• USA Home
  • 900037 - Iron oxide (II,III), nanoparticles

EMAIL THIS PAGE TO A FRIEND
900037 Sigma-Aldrich

Iron oxide (II,III), nanoparticles

15 nm avg. part. size (TMS), 1 mg/mL (in 10 mM PBS buffer), biotin functionalized

Purchase

Properties

Related Categories 26: Fe, Biomedical Materials, Materials Science, Nanomaterials, Nanoparticles: Materials by Application,
Quality Level   100
form   dispersion
concentration   1 mg/mL (in 10 mM PBS buffer)
avg. part. size   15 nm (TMS)
functional group   biotin
storage temp.   2-8°C

Description

Application

Iron oxide nanoparticles exhibit biocompatibility and non-toxicity and are used as magnetic resonance imaging contrast agents, gene carriers for gene therapy, therapeutic agents for hyperthermia-based cancer treatments, in magnetic sensing probes for in vivo diagnostics and target specific drug delivery.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

Recent Developements in Magnetic Iron Oxide Nanoparticles for Non-Invasive Bioimaging

1 Department of Radiology and Imaging Sciences, Emory University School of Medicine, Atlanta, Georgia 30322, USA 2 Ocean Nanotech, LLC, 7964 Arjons Drive, San Diego, California 92126, USA *E-mail: hm...
Keywords: Absorption, Clinical, Diagnostic, Diagnostic Imaging, Diseases, Electron spin resonance, Growth factors, Infrared spectroscopy, Ligands, Magnetic resonance imaging, Microscopy, Microwave synthesis, Nanomaterials, Positron Emission Tomography, Scanning electron microscopy, Transmission electron microscopy

Synthesis and Application of Shape-Controlled Fe3O4 Nanostructures

Magnetite (Fe3O4) nanocrystals have extensive applications in drug and gene delivery,1 field-directed assembly,2 energy storage,3 and catalysis4 due to their excellent magnetic properties, ready avai...
Keywords: Absorption, Cancer, Diffraction, Ligands, Magnetic resonance imaging, Microscopy, Nanomaterials, Nucleic acid annealing, Reductions, Scanning electron microscopy, Solvents, Sonication, Transmission electron microscopy, transformation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?