• USA Home
  • 900350 - Ultroxa®: Poly(2-methyl-2-oxazoline)

EMAIL THIS PAGE TO A FRIEND
900350 Aldrich

Ultroxa®: Poly(2-methyl-2-oxazoline)

average Mn 10,000, PDI ≤1.2

Synonym: Me-PMeOx-OH

  • Linear Formula CH3(C4H7NO)nOH

Purchase

Description

Packaging

1 g in glass bottle

Legal Information

Ultroxa is a registered trademark of University of Ghent

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Poly(2-Oxazoline)S: The Versatile Polymer Platform for Biomedicine

Victor R. de la Rosa,1 An Van Den Bulcke,2 and Richard Hoogenboom1* 1Supramolecular Chemistry Group, Department of Organic and Macromolecular Chemistry Ghent University, Krijgslaan 281-S4, 9000 Ghent...
Victor R. de la Rosa, An Van Den Bulcke, and Richard Hoogenboom
Material Matters, 2016, 11.3
Keywords: Adhesion, Adsorption, Amidations, Bacterial conjugations, Biomaterials, Cancer, Catalysis, Click chemistry, Clinical, Cosmetics, Cycloadditions, Degradations, Electron spin resonance, Pharmaceutical, Polymerization reactions, Ring-opening polymerization

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?


Webinar - Hydrogels