EMAIL THIS PAGE TO A FRIEND
900466 Sigma-Aldrich

Gold nanoparticles

70 nm, maleimide functionalized, conjugation kit

Synonym: Au nanoparticles

  •  NACRES NA.23

Purchase

Properties

Related Categories 79: Au, Biomedical Materials, Gold Nanoparticle Kits, Gold Nanoparticles, Materials Science,
form   nanoparticles
shelf life   12 mo. at -20 °C ( If stored unopened and as specified, are stable for at least 12 months.)
packaging   pkg of 1 ea
storage condition   protect from light
particle size   70 nm
λmax   548 nm
functional group   maleimide
storage temp.   −20°C

Description

Application

Biocompatibility, facile conjugation to biomolecules and unique optical properties arising from surface plasmon resonance render the nanoparticles of gold useful for variety of biomedical research such as bio-imaging, bio-sensing, photothermal hyperthermia, drug delivery etc. Our maleimide functionalized gold nanoparticles kits contain ready-to-use mixtures and have been optimized for high efficiency one-step conjugation of thiol-modified ligands such as oligonucleotides, antibodies, antibody fragments, proteins and peptides.

These kits contain the following components:
1. Maleimide gold nanoparticles (lyophilized).
2. Ligand resuspension buffer.
3. Reaction buffer.
4. Quencher (lyophilized).

Legal Information

Product of CytoDiagnostics, Inc.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Increasing the Sensitivity of Lateral Flow Diagnostic Assays with Ultra-Bright Gold Nanoshell Reporters

Lateral flow assays (also known as immunochromatographic assays) detect or quantify biomolecules in complex samples including blood, urine, saliva, or other fluids to diagnose a variety of medical co...
Keywords: Absorption, Adsorption, Bacterial conjugations, Centrifugation, Deposition, Detergents, Diagnostic, Filtration, Flow cytometry, Hormones, Hydration reaction, Immobilization, Ligands, Microscopy, Raman spectroscopy, Scanning electron microscopy, Spectroscopy, Transmission electron microscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?