EMAIL THIS PAGE TO A FRIEND
900782 Sigma-Aldrich

SF-PDI

  • CAS Number 1643842-69-6

  • Empirical Formula (Hill Notation) C125H136N4O8

  • Molecular Weight 1822.44

Purchase

Properties

Related Categories Materials Science, Non-fullerene Acceptor Materials, Organic Field Effect Transistor (OFET) Materials, Organic Photovoltaic (OPV) Materials, Organic and Printed Electronics,
assay   99%
form   solid
solubility   chlorobenzene: soluble
  chloroform: soluble
  dichlorobenzene: soluble

Description

Packaging

100 mg in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Organic Photovoltaic Applications for IoT, Architecture, and Wearables

1Merck Chemicals Ltd, Chilworth Technical Centre, University Parkway, Southampton. SO16 7QD, U.K. 2Merck KGaA, Frankfurter Str. 250, 64293 Darmstadt I Germany 3Merck Selbstmedikation GmbH, Frankfurte...
Graham Morse,1 Richard Harding,1 Nicolas Blouin,2 Hannah Buerckstrummer2
Not pictured: Agnieszka Pron,1 Stephan Wieder,2 David Mueller,2 and Stephane Berny3
Material Matters, 2017, 12.3
Keywords: Absorption, Alternative energy, Catalysis, Coupling reactions, Cross couplings, Deposition, Environmental, Indicators, Industries, PAGE, Photovoltaics, Polymerization reactions, Positron Emission Tomography, Purification, Safety industry, Semiconductor, Separation, Solar cells, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?