• USA Home
  • 900838 - 1-Ethynyl-4-(1,2,2-triphenylethenyl)benzene

EMAIL THIS PAGE TO A FRIEND
900838 Sigma-Aldrich

1-Ethynyl-4-(1,2,2-triphenylethenyl)benzene

Synonym: 1-(4-Ethylnylphenyl)-1,2,2-triphenylethene, [4-(1,2,2-Triphenylvinyl)phenyl]acetylene

  • CAS Number 1225493-18-4

  • Empirical Formula (Hill Notation) C28H20

  • Molecular Weight 356.46

Purchase

Description

Application

AIE (aggregation-induced emission)-active building block.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?