EMAIL THIS PAGE TO A FRIEND
900897 Sigma-Aldrich

Hyflon® AD 40H SF

high purity

Synonym: Amorphous perfluoropolymer solution, P(TTD-TFE), Poly(tetrafluoroethylene-co-2,2,4-trifluoro-5-trifluoromethoxy-1,3-dioxole), TTD-TFE copolymers

  • CAS Number 161611-79-6

  • Linear Formula (C4F6O3)n(C2F4)m

Purchase

Properties

Related Categories Actuators, Bioelectronics, Ferroelectric Polymers, Flexible Organic Transistors, Fluorocarbons,
description   Polymer content: Hyflon® AD 40H 2.0 ± 0.5 wt%
  Solution in perfluorinated solvent (Galden® D02TS)
composition   TTD content, 40 ±5 mol % by
transmittance   480-800 nm, >99%
viscosity   80 cP(25 °C, Brookfield)

Description

General description

Hyflon® AD 40H SF is a high purity Hyflon® AD 40H perfluoropolymer solution in a perfluorinated solvent. This can be considered ready-to-use solution suitable for application in the most demanding solution process technologies.
• Amorphous glassy polymer structure.
• Solubility in perfluorinated solvents.
• Low dielectric constant of 2.0 at 1 kHz.

Excellent optical properties:
• High UV transmittance.
• Low refractive index.

Hyflon® AD can provide uniform and thin films or coatings (down to submicron thickness if required) by casting or coating techniques and thermal treatments, well below the typical baking temperatures of semicrystalline perflouropolymers.

Key features of Hyflon® AD amorphous perfluoropolymers:
• High temperature stability.
• Good chemical resistance.
• Excellent hydrophobicity.
• Very low surface energy.
• Low dielectric constant.
• Very high gas permeability and selectivity.
• Outstanding optical properties (e.g. high UV transmittance, low refractive index).

Examples of typical applications include:
• Protective coatings.
• Interlayer dielectric films and pellicles for electronics.

Legal Information

Hyflon is a registered trademark of Solvay Group

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?