EMAIL THIS PAGE TO A FRIEND
900902 Sigma-Aldrich

Hyflon® AD 40H

Synonym: Amorphous perfluoropolymer, P(TTD-TFE), Poly(tetrafluoroethylene-co-2,2,4-trifluoro-5-trifluoromethoxy-1,3-dioxole), TTD-TFE copolymers

  • CAS Number 161611-79-6

  • Linear Formula (C4F6O3)n(C2F4)m

Purchase

Properties

Related Categories Actuators, Bioelectronics, Ferroelectric Polymers, Flexible Organic Transistors, Fluorocarbons,
description   Glass Transition Temperature: 90 °C (ASTM D3418)
  Specific gravity: 1.98 (ASTM D792)
form   powder
composition   TTD content, 40 mol % ±5 mol % by
refractive index   n/D 1.331 (ASTM D542)
intrinsic viscosity   120 cm3/g±30 cm3/g(30 °C) (ASTM D2857)

Description

Application

Hyflon® AD 40H is an amorphous perfluoropolymer available in powder form.

• Amorphous glassy polymer structure.
• Solubility in perfluorinated solvents.
• Low dielectric constant of 2.0 at 1 kHz.
• Excellent optical properties:
- High UV transmittance.
- Low refractive index.

Hyflon® AD can provide uniform and thin films or coatings (down to submicron thickness if required) by casting or coating techniques and thermal treatments, well below the typical baking temperatures of semicrystalline perfluoropolymers.

Key features of Hyflon® AD amorphous perfluoropolymers:
• High temperature stability.
• Good chemical resistance.
• Excellent hydrophobicity.
• Very low surface energy.
• Low dielectric constant.
• Very high gas permeability and selectivity.
• Outstanding optical properties (e.g. high UV transmittance, low refractive index).

Examples of typical applications include:
• Protective coatings.
• Interlayer dielectric films and pellicles for electronics.

Legal Information

Hyflon is a registered trademark of Solvay Group

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?