• USA Home
  • 900952 - 2-[(1E,3E)-6-[3-(Dimethylamino)pyridine]-1,3-butadien-1-yl]-4,5-dihydro-4-thiazolecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
900952 Sigma-Aldrich

2-[(1E,3E)-6-[3-(Dimethylamino)pyridine]-1,3-butadien-1-yl]-4,5-dihydro-4-thiazolecarboxylic acid

Synonym: DP3, TokeOni analogue, Water-soluble NIR emission luciferin analogue, seMpai

  • Empirical Formula (Hill Notation) C15H17N3O2S

  • Molecular Weight 303.38

Purchase

Properties

Related Categories Infrared (IR) Dyes, Materials Science, Organic and Printed Electronics, Photonic and Optical Materials
Quality Level   100
form   powder
storage temp.   −20°C
SMILES string   CN(C)C1=CN=C(/C=C/C=C/C2=NC(C(O)=O)CS2)C=C1

Description

General description

Luciferin is a generic term for the light-emitting compound found in organisms that generate bioluminescence.
This product is a new NIR emission luciferin analogue having luminescence peak at 670~680 nm with a peak range in the near-infrared (NIR) region. It has the enhanced solubility of 20 mg/mL compared to HCl free form of TokeOni (0.2 mg/mL).

Application

• Bio imaging.
• In vivo imaging.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Synthesis and Applications of Graphene Nanoribbons Synthesized

1 Department of Chemistry, 2 Smalley-Curl Institute and The NanoCarbon Center, 3 Department of Materials Science and NanoEngineering, Rice University, 6100 Main Street MS 222, Houston, Texas 77005, U...
Keywords: Building blocks, Capillary electrophoresis, Mass spectrometry, Materials Science, Nanotubes, Nucleic acid hybridization, Oxidations, Phase transitions, Polymerization reactions, Sequencing, Solvents

TokeOni Analog - seMpai: Properties and Applications

Bioluminescence imaging (BLI) has been an indispensable technique for noninvasive and highly sensitive observation of biological events in living tissues1-3. Firefly luciferase (Fluc) and D-luciferin...
Takahiro Kuchimaru

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?