EMAIL THIS PAGE TO A FRIEND
900959 Sigma-Aldrich

Bepp2

sublimed

Synonym: Bis(2-(2-hydroxyphenyl)pyridinato)beryllium, Bis(2-(2-hydroxyphenyl)pyridine)beryllum, Bis[2-(2-pyridinyl)phenolato]beryllium

  • CAS Number 220694-90-6

  • Empirical Formula (Hill Notation) C22H16BeN2O2

  • Molecular Weight 349.39

Purchase

Properties

Related Categories Light Emitters and Dopants, Materials Science, OLED and PLED Materials, Organic and Printed Electronics
assay   99% (sublimed)
form   solid

Description

General description

Blue OLED dopant material.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Organic Optoelectronics on Shape Memory Polymers

Center for Organic Photonics and Electronics (COPE), School of Electrical and Computer Engineering, Georgia Institute of Technology, Atlanta, Georgia 30332, USA *E-mail: bernard.kippelen@ece.gatech.e...
Canek Fuentes-Hernandez, Bernard Kippelen*
Material Matters, 2017, 12.3
Keywords: AGE, Adhesion, Atomic layer deposition, Biological processes, Catalysis, Ceramics, Deposition, Electronics, Environmental, Nanomaterials, Nanotubes, Oxidations, Photovoltaics, Polymerization reactions, Reductions, Semiconductor, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?