EMAIL THIS PAGE TO A FRIEND
901045 Sigma-Aldrich

J61

Synonym: Poly[[5,6-difluoro-2-(2-hexyldecyl)-2H-benzotriazole-4,7-diyl]-2,5-thiophenediyl[4,8-bis[5-(dodecylthio)-2-thienyl]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl]-2,5-thiophenediyl]

Purchase

Properties

mol wt   Mw 50,000-100,000 by GPC (PS standard)
solubility   organic solvents: soluble (Soluble in chlorobenzene and dichlorobenzene,Limited solubility in CHCl3)
PDI   2.0‑3.0
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles

Articles

Organic Photovoltaic Applications for IoT, Architecture, and Wearables

1Merck Chemicals Ltd, Chilworth Technical Centre, University Parkway, Southampton. SO16 7QD, U.K. 2Merck KGaA, Frankfurter Str. 250, 64293 Darmstadt I Germany 3Merck Selbstmedikation GmbH, Frankfurte...
Graham Morse,1 Richard Harding,1 Nicolas Blouin,2 Hannah Buerckstrummer2
Not pictured: Agnieszka Pron,1 Stephan Wieder,2 David Mueller,2 and Stephane Berny3
Material Matters, 2017, 12.3
Keywords: Absorption, Alternative energy, Catalysis, Coupling reactions, Cross couplings, Deposition, Environmental, Indicators, Industries, PAGE, Photovoltaics, Polymerization reactions, Positron Emission Tomography, Purification, Safety industry, Semiconductor, Separation, Solar cells, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?