• USA Home
  • 901405 - Poly(ethylene glycol) linear dendron

EMAIL THIS PAGE TO A FRIEND
901405 Sigma-Aldrich

Poly(ethylene glycol) linear dendron

NHBoc terminated, PEG Mn 2000, generation 4

Synonym: Dendron functionalized PEG, PFD-G4-PEG2k-NHBOC

  • Empirical Formula (Hill Notation) C294H512N16O139

  • Molecular Weight 6495.23

  •  NACRES NA.23

Purchase

Properties

Related Categories Amine-terminated PEG-core Dendrimers, Dendrimers, Materials Science, Nanomaterials
Quality Level   100
form   solid
color   white to off-white
storage temp.   −20°C
SMILES string   CC(COC(CCNC(OC(C)(C)C)=O)=O)(C(OCC(C(OCC(C(OCC(C(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)(C)COC(C(COC(C(COC(C(COC(CCNC(OC(C)(C)C)=O)=O)(C)COC(CCNC(OC(C)(C

Description

General description

• Polymer architecture: PEG-linear dendron
• Generation: 4
• PEG Average Mn: 2,000
• End Group Functionality: Boc-protected amine
• No. Surface Groups: 16
• Calculated Mol. Wt.: 6479.30 g/mol
• Boc : tert-Butyloxycarbonyl
• Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid

Application

Bis-MPA (or 2,2-Bis(methylol)propionic acid) is an aliphatic, pro-chiral molecule comprised of two hydroxyls and one carboxylic group that has been used in the synthesis of many different types of polymers. Dendrons synthesized from Bis-MPA are biodegradable and have low cytotoxicity, making them well suited for use in biological research applications. These materials have been used in many applications such as signal amplification in bioassays and in drug delivery applications. PEG-core dendrons feature excellent solubility in water and a high degree of functionality per PEG chain, making them strong candidates for advanced drug delivery systems or as targeting vessels. Additionally, the terminal amine groups can readily be used in EDC or DCC coupling reactions (after Boc deprotection) with carbonyl-containing compounds, to yield highly functionalized materials for a variety of biomedical applications.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?