EMAIL THIS PAGE TO A FRIEND
90205 Sigma-Aldrich

DL-β-Homomethionine

≥99.0% (TLC)

Synonym: (±)-3-Amino-5-(methylthio)pentanoic acid

  • CAS Number 158570-14-0

  • Linear Formula CH3SCH2CH2CH(NH2)CH2COOH

  • Molecular Weight 163.24

  •  Beilstein/REAXYS Number 6591767

  •  MDL number MFCD06205952

  •  PubChem Substance ID 57653375

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
Quality Level   100
assay   ≥99.0% (TLC)
application(s)   peptide synthesis: suitable
mp   190-193 °C (dec.)
SMILES string   CSCCC(N)CC(O)=O
InChI   1S/C6H13NO2S/c1-10-3-2-5(7)4-6(8)9/h5H,2-4,7H2,1H3,(H,8,9)
InChI key   QWVNCDVONVDGDV-UHFFFAOYSA-N

Description

Packaging

1 g in poly tube

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?