• USA Home
  • 91285 - Trichloromethanesulfinyl chloride

EMAIL THIS PAGE TO A FRIEND
91285 Aldrich

Trichloromethanesulfinyl chloride

technical, ≥85% (AT)

Purchase

Properties

grade   technical
assay   ≥85% (AT)
refractive index   n20/D 1.541
bp   38-40 °C/2 mmHg(lit.)
density   1.750 g/mL at 20 °C(lit.)

Description

Other Notes

For the generation of dichlorocarbene

Safety & Documentation
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?