• USA Home
  • 91724 - Ethyl (R)-2-(trifluoromethylsulfonyloxy)propionate

EMAIL THIS PAGE TO A FRIEND
91724 Sigma-Aldrich

Ethyl (R)-2-(trifluoromethylsulfonyloxy)propionate

≥97.0%

Synonym: Ethyl O-trifluoromethanesulfonyl-D-lactate, Ethyl D-lactate 2-O-triflate

  • CAS Number 84028-89-7

  • Empirical Formula (Hill Notation) C6H9F3O5S

  • Molecular Weight 250.19

  •  Beilstein/REAXYS Number 5271893

  •  MDL number MFCD00674552

  •  PubChem Substance ID 57653451

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters, Organic Building Blocks More...
Quality Level   100
assay   ≥97.0%
optical activity   [α]20/D +42±2°, neat
contains   ~1% magnesium oxide as stabilizer (white solid)
refractive index   n20/D 1.375
density   1.342 g/mL at 20 °C (lit.)
storage temp.   −20°C
SMILES string   CCOC(=O)[C@@H](C)OS(=O)(=O)C(F)(F)F
InChI   1S/C6H9F3O5S/c1-3-13-5(10)4(2)14-15(11,12)6(7,8)9/h4H,3H2,1-2H3/t4-/m1/s1
InChI key   RYSBPHXTNVWICH-SCSAIBSYSA-N

Description

Packaging

5 mL in glass bottle

Caution

may discolor to brown on storage

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?