• USA Home
  • 91942 - 2,4,6-Trihydroxybenzoic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
91942 Sigma-Aldrich

2,4,6-Trihydroxybenzoic acid monohydrate

technical, ≥90% (T)

Synonym: Phloroglucinolcarboxylic acid

  • CAS Number 71989-93-0

  • Empirical Formula (Hill Notation) C7H6O5 · H2O

  • Molecular Weight 188.13

  •  Beilstein/REAXYS Number 2212148

  •  EC Number 201-467-5

  •  MDL number MFCD00149095

  •  PubChem Substance ID 57653461

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   HWZIRFCGHAROOI-UHFFFAOYSA-N
grade   technical
assay   ≥90% (T)
ign. residue   ~0.5%
mp   205 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DH8910000

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

367346 2,4,6-Trihydroxybenzoic acid monohydrate, technical grade, 90%, predominantly 1,3,5-benzenetriol

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?