• USA Home
  • 92282 - (R)-3-(Fmoc-amino)-5-phenyl-pentanoic acid

EMAIL THIS PAGE TO A FRIEND
92282 Sigma-Aldrich

(R)-3-(Fmoc-amino)-5-phenyl-pentanoic acid

≥95.0%

Synonym: Fmoc-5-phenyl-D-β-norvaline

Purchase

Properties

assay   ≥95.0%
storage temp.   2-8°C
SMILES string   OC(=O)C[C@@H](CCc1ccccc1)NC(=O)OCC2c3ccccc3-c4ccccc24
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?