• USA Home
  • 93669 - (R)-4-(Fmoc-amino)-5-phenylpentanoic acid

EMAIL THIS PAGE TO A FRIEND
93669 Sigma-Aldrich

(R)-4-(Fmoc-amino)-5-phenylpentanoic acid

≥97.0%

Synonym: (R)-4-(Fmoc-amino)-5-phenylvaleric acid, Fmoc-γ-L-dihomo-phenylalanine

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Other Unnatural Amino Acids & Derivatives, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   ≥97.0%
form   powder with small lumps
application(s)   peptide synthesis: suitable
color   white to faintly brown
functional group   Fmoc
storage temp.   2-8°C
SMILES string   OC(=O)CC[C@H](Cc1ccccc1)NC(=O)OCC2c3ccccc3-c4ccccc24
InChI   1S/C26H25NO4/c28-25(29)15-14-19(16-18-8-2-1-3-9-18)27-26(30)31-17-24-22-12-6-4-10-20(22)21-11-5-7-13-23(21)24/h1-13,19,24H,14-17H2,(H,27,30)(H,28,29)/t19-/m1/s1
InChI key   GQTVSSUIIKDXQC-LJQANCHMSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?