• USA Home
  • 94557 - (S)-Boc-4-methyl-β-Homophe-OH

EMAIL THIS PAGE TO A FRIEND
94557 Sigma-Aldrich

(S)-Boc-4-methyl-β-Homophe-OH

≥97.0% (HPLC)

Synonym: (S)-3-(Boc-amino)-4-(4-methylphenyl)butyric acid, Boc-4-methyl-L-β-homophenylalanine

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
InChI Key   KAHQBNUMNOJEPZ-ZDUSSCGKSA-N
assay   ≥97.0% (HPLC)

Description

Packaging

500 mg in poly tube

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?