• USA Home
  • 94603 - (S)-Boc-3-(trifluoromethyl)-β-Homophe-OH

EMAIL THIS PAGE TO A FRIEND
94603 Sigma-Aldrich

(S)-Boc-3-(trifluoromethyl)-β-Homophe-OH

≥98.0% (HPLC)

Synonym: (S)-3-(Boc-amino)-4-[3-(trifluoromethyl)phenyl]butyric acid, Boc-3-(trifluoromethyl)-L-β-homophenylalanine

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   ≥98.0% (HPLC)
application(s)   peptide synthesis: suitable
storage temp.   room temp
SMILES string   CC(C)(C)OC(=O)N[C@H](CC(O)=O)Cc1cccc(c1)C(F)(F)F
InChI   1S/C16H20F3NO4/c1-15(2,3)24-14(23)20-12(9-13(21)22)8-10-5-4-6-11(7-10)16(17,18)19/h4-7,12H,8-9H2,1-3H3,(H,20,23)(H,21,22)/t12-/m0/s1
InChI key   ATUJVGXTABSYTK-LBPRGKRZSA-N

Description

Packaging

500 mg in poly tube

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?