• USA Home
  • 94884 - 1,1,1,2,3,4,4,5,5,5-Decafluoropentane

EMAIL THIS PAGE TO A FRIEND
94884 Sigma-Aldrich

1,1,1,2,3,4,4,5,5,5-Decafluoropentane

technical, ~60% (GC)

Synonym: 2H,3H-Perfluoropentane, Vertrel XF

  • CAS Number 138495-42-8

  • Linear Formula CF3CFHCFHCF2CF3

  • Molecular Weight 252.05

  •  Beilstein/REAXYS Number 6324333

  •  MDL number MFCD00798129

  •  PubChem Substance ID 57653646

  •  NACRES NA.22

Purchase

Properties

Related Categories Alkyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   RIQRGMUSBYGDBL-UHFFFAOYSA-N
grade   technical
assay   ~60% (GC)
impurities   other fluoropentanes, residual
refractive index   n20/D 1.3
bp   55 °C (lit.)
density   1.60 g/mL at 20 °C (lit.)

Description

Application

1,1,1,2,3,4,4,5,5,5-Decafluoropentane (2H,3H-perfluoropentane or Vertrel XF or DFP) can be used as a solvent in the Jacobsen-Katsuki epoxidation of styrene using 2-phenylpyridine N-oxide as donor ligand. It can be also used as a grinding liquid in mechanosynthesis of double metal cyanide complexes applicable as catalysts for co-polymerization of CO2 and propylene oxide.

Packaging

100, 500 mL in glass bottle

Safety & Documentation

Safety Information

Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
RZ9885000
Flash Point(F) 
>257 °F
Flash Point(C) 
>125 °C
Protocols & Articles

Articles

Solvents for Fluorous Synthesis

Following the definition offered by Gladysz and Curran[1] a “Fluorous Medium” is “any phase of a perfluoroalkane, perfluorodialkylether, perfluorotrialkylamine, or similar non-polar species, or any s...
ChemFiles Volume 4 Article 9
Keywords: Methods, Separation, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?