• USA Home
  • 95516 - 4-Piperazinobenzonitrile

EMAIL THIS PAGE TO A FRIEND
95516 Sigma-Aldrich

4-Piperazinobenzonitrile

≥97.0%

Synonym: 1-(4-Cyanophenyl)piperazine

  • CAS Number 68104-63-2

  • Empirical Formula (Hill Notation) C11H13N3

  • Molecular Weight 187.24

  •  Beilstein/REAXYS Number 783828

  •  MDL number MFCD01320890

  •  PubChem Substance ID 57653683

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
assay   ≥97.0%
mp   83-86 °C
SMILES string   N#Cc1ccc(cc1)N2CCNCC2
InChI   1S/C11H13N3/c12-9-10-1-3-11(4-2-10)14-7-5-13-6-8-14/h1-4,13H,5-8H2
InChI key   DJJNYEXRPRQXPD-UHFFFAOYSA-N

Description

Packaging

1 g in poly tube

5 g in poly bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?